Assay Details
| Gene Symbol: BCL7C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265776.2 | NP_001252705.1 | 638 | 87 | ||
| XM_028840558.1 | XP_028696391.1 | 1524 | 87 | |||
| GenBank mRNA | JU327130.1 | - | 484 | 87 | ||
| JV566314.1 | - | 593 | 87 | |||
| JV635665.1 | - | 638 | 87 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

BCL7C
Genomic Map