Assay Details
| Gene Symbol: SNAP29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261361.2 | NP_001248290.1 | 706 | 85 | ||
| GenBank mRNA | JU337134.1 | - | 85 | |||
| JU473624.1 | - | 85 | ||||
| JV044934.1 | - | 706 | 85 | |||
| JV638698.1 | - | 707 | 85 | |||
More Information
Set Membership: |
|

SNAP29
Genomic Map