Assay Details
| Gene Symbol: HOMER2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015142878.2 | XP_014998364.2 | 816 | 135 | ||
| XM_028851260.1 | XP_028707093.1 | 816 | 135 | |||
| XM_028851261.1 | XP_028707094.1 | 816 | 135 | |||
| XM_028851262.1 | XP_028707095.1 | 936 | 135 | |||
| XM_028851263.1 | XP_028707096.1 | 876 | 135 | |||
| XM_028851264.1 | XP_028707097.1 | 841 | 135 | |||
| XM_028851265.1 | XP_028707098.1 | 1505 | 135 | |||
| XM_028851266.1 | XP_028707099.1 | 786 | 135 | |||
| GenBank mRNA | JU337185.1 | - | 766 | 135 | ||
| JV598870.1 | - | 789 | 135 | |||
More Information
Set Membership: |
|

HOMER2
Genomic Map