Assay Details
Gene Symbol: CDX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001095271.2 | XP_001095271.1 | 802 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
