Assay Details
| Gene Symbol: FOXN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015144359.2 | XP_014999845.1 | 1029 | 105 | ||
| XM_015144360.2 | XP_014999846.1 | 999 | 105 | |||
| XM_015144361.2 | XP_014999847.1 | 996 | 105 | |||
| XM_028851735.1 | XP_028707568.1 | 990 | 105 | |||
| XM_028851736.1 | XP_028707569.1 | 996 | 105 | |||
| GenBank mRNA | JU332703.1 | - | 752 | 105 | ||
| JV046053.1 | - | 774 | 105 | |||
More Information
Set Membership: |
|

FOXN3
Genomic Map