Assay Details
| Gene Symbol: LOXL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015142675.2 | XP_014998161.1 | 2007 | 70 | ||
| XM_028851107.1 | XP_028706940.1 | 206 | 70 | |||
| GenBank mRNA | JU326682.1 | - | 194 | 70 | ||
| JU486585.1 | - | 224 | 70 | |||
| JV651375.1 | - | 402 | 70 | |||
More Information
Set Membership: |
|

LOXL1
Genomic Map