Assay Details
| Gene Symbol: GLYAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257270.1 | NP_001244199.1 | 181 | 159 | ||
| GenBank mRNA | CO583152.1 | - | 190 | 159 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001257270.1 transcripts.
Set Membership: |
|

GLYAT
Genomic Map