Assay Details
| Gene Symbol: GLYAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257270.1 | NP_001244199.1 | 427 | 103 | ||
| XM_015114098.2 | XP_014969584.2 | 491 | 103 | |||
| GenBank mRNA | CK231271.1 | - | 271 | 103 | ||
| CO583152.1 | - | 436 | 103 | |||
| JU478673.1 | - | 278 | 103 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

GLYAT
Genomic Map