Assay Details
| Gene Symbol: RAMP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257448.1 | NP_001244377.1 | 248 | 115 | ||
| XM_028844823.1 | XP_028700656.1 | 560 | 115 | |||
| GenBank mRNA | JU332466.1 | - | 248 | 115 | ||
More Information
Set Membership: |
|

RAMP3
Genomic Map