Assay Details
Gene Symbol: ENDOU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015151532.2 | XP_015007018.2 | 6769 | 86 | ||
XM_015151533.2 | XP_015007019.2 | 1410 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
