Assay Details
| Gene Symbol: PNLIPRP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028826903.1 | XP_028682736.1 | 1137 | 94 | ||
| XM_028826904.1 | XP_028682737.1 | 1046 | 94 | |||
More Information
Set Membership: |
|

PNLIPRP1
Genomic Map