Assay Details
| Gene Symbol: S100A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015109628.2 | XP_014965114.1 | 316 | 94 | ||
| GenBank mRNA | JU473820.1 | - | 260 | 94 | ||
| JV045809.1 | - | 178 | 94 | |||
| JV477929.1 | - | 63 | 94 | |||
| JV567565.1 | - | 265 | 94 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

S100A1
Genomic Map