Assay Details
Gene Symbol: TIMM23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190973.2 | NP_001177902.1 | 641 | 119 | ||
XM_028853456.1 | XP_028709289.1 | 626 | 119 | |||
GenBank mRNA | JU320757.1 | - | 640 | 119 | ||
JU471088.1 | - | 646 | 119 | |||
JV044525.1 | - | 644 | 119 | |||
JV491241.1 | - | 560 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
