Assay Details
| Gene Symbol: SLCO1B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028829310.1 | XP_028685143.1 | 2003 | 80 | ||
| XM_028829311.1 | XP_028685144.1 | 2003 | 80 | |||
| GenBank mRNA | JV705369.1 | - | 1679 | 80 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SLCO1B1
Genomic Map