Assay Details
| Gene Symbol: RASL10A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260903.2 | NP_001247832.1 | 708 | 106 | ||
| GenBank mRNA | JU322310.1 | - | 225 | 106 | ||
| JV603803.1 | - | 240 | 106 | |||
| JV667618.1 | - | 706 | 106 | |||
| JV670866.1 | - | 706 | 106 | |||
More Information
Set Membership: |
|

RASL10A
Genomic Map