Assay Details
| Gene Symbol: LGALS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001087063.4 | XP_001087063.1 | 329 | 91 | ||
| XM_028827743.1 | XP_028683576.1 | 272 | 91 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LGALS2| Gene Symbol: LGALS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001087063.4 | XP_001087063.1 | 329 | 91 | ||
| XM_028827743.1 | XP_028683576.1 | 272 | 91 | |||
Set Membership: |
|