Assay Details
| Gene Symbol: CCL27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032876.1 | NP_001028048.1 | 206 | 146 | ||
| GenBank mRNA | AF449278.1 | - | 206 | 146 | ||
| JV640692.1 | - | 233 | 146 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CCL27
Genomic Map