Assay Details
| Gene Symbol: LOC705843 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015144470.2 | XP_014999956.1 | 1222 | 106 | ||
| XM_028851810.1 | XP_028707643.1 | 1141 | 106 | |||
| XM_028851811.1 | XP_028707644.1 | 985 | 106 | |||
| GenBank mRNA | JU327533.1 | - | 979 | 106 | ||
| JV531498.1 | - | 983 | 106 | |||
| JV667329.1 | - | 989 | 106 | |||
More Information
Set Membership: |
|

LOC705843
Genomic Map