Assay Details
Gene Symbol: SUGT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001084516.4 | XP_001084516.2 | 485 | 154 | ||
XM_015121145.2 | XP_014976631.2 | 583 | 154 | |||
XM_028837586.1 | XP_028693419.1 | 1861 | 154 | |||
GenBank mRNA | JU321391.1 | - | 491 | 154 | ||
JU321392.1 | - | 492 | 154 | |||
JU471332.1 | - | 476 | 154 | |||
JV045708.1 | - | 476 | 154 | |||
JV637908.1 | - | 535 | 154 |

More Information
Set Membership: |
![]() ![]() ![]() |
