Assay Details
Gene Symbol: FOSB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015124278.2 | XP_014979764.1 | 1048 | 69 | ||
XM_028839678.1 | XP_028695511.1 | 931 | 69 | |||
XM_028839679.1 | XP_028695512.1 | 2806 | 69 | |||
GenBank mRNA | JV611420.1 | - | 368 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
