Assay Details
| Gene Symbol: KDELR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261069.1 | NP_001247998.1 | 382 | 150 | ||
| XM_028838489.1 | XP_028694322.1 | 200 | 150 | |||
| GenBank mRNA | JU332650.1 | - | 384 | 150 | ||
| JU474093.1 | - | 382 | 150 | |||
| JV492778.1 | - | 234 | 150 | |||
| Gene Symbol: KDELR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JV046035.1 | - | 387 | 150 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

KDELR1
Genomic Map