Assay Details
| Gene Symbol: ZNF41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015127292.2 | XP_014982778.1 | 634 | 148 | ||
| XM_015127294.2 | XP_014982780.1 | 618 | 148 | |||
| XM_028842512.1 | XP_028698345.1 | 3648 | 148 | |||
| GenBank mRNA | JU336287.1 | - | 417 | 148 | ||
| JV636363.1 | - | 327 | 148 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028842512.1 transcripts.
Set Membership: |
|

ZNF41
Genomic Map