Assay Details
| Gene Symbol: CHP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266371.1 | NP_001253300.1 | 278 | 66 | ||
| GenBank mRNA | JU320186.1 | - | 283 | 66 | ||
| JU474170.1 | - | 159 | 66 | |||
| JV045394.1 | - | 278 | 66 | |||
| JV641251.1 | - | 285 | 66 | |||
More Information
Set Membership: |
|

CHP1
Genomic Map