Assay Details
Gene Symbol: GNA12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015133200.2 | XP_014988686.1 | 694 | 92 | ||
XM_028845522.1 | XP_028701355.1 | 317 | 92 | |||
XM_028845523.1 | XP_028701356.1 | 400 | 92 | |||
GenBank mRNA | JU327438.1 | - | 451 | 92 | ||
JV465160.1 | - | 444 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
