Assay Details
Gene Symbol: CASC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260566.1 | NP_001247495.1 | 650 | 82 | ||
XM_015119568.2 | XP_014975054.1 | 1863 | 82 | |||
GenBank mRNA | JU333443.1 | - | 666 | 82 | ||
JV046462.1 | - | 650 | 82 | |||
JV609123.1 | - | 673 | 82 | |||
JV636033.1 | - | 656 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
