Assay Details
| Gene Symbol: TMED3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257920.1 | NP_001244849.1 | 217 | 70 | ||
| GenBank mRNA | JU333688.1 | - | 217 | 70 | ||
| JU474934.1 | - | 280 | 70 | |||
| JV046591.1 | - | 222 | 70 | |||
| JV646856.1 | - | 281 | 70 | |||
More Information
Set Membership: |
|

TMED3
Genomic Map