Assay Details
| Gene Symbol: TMED3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257920.1 | NP_001244849.1 | 467 | 123 | ||
| GenBank mRNA | JU333688.1 | - | 467 | 123 | ||
| JU474934.1 | - | 530 | 123 | |||
| JV046591.1 | - | 472 | 123 | |||
| JV646856.1 | - | 531 | 123 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TMED3
Genomic Map