Assay Details
Gene Symbol: LOC114675597 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028842817.1 | XP_028698650.1 | 358 | 119 |
Gene Symbol: CASP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001111598.4 | XP_001111598.2 | 703 | 119 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
