Assay Details
| Gene Symbol: SLC25A10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267019.1 | NP_001253948.1 | 506 | 85 | ||
| XM_015120547.2 | XP_014976033.1 | 493 | 85 | |||
| XM_028835659.1 | XP_028691492.1 | 588 | 85 | |||
| XM_028835660.1 | XP_028691493.1 | 1039 | 85 | |||
| GenBank mRNA | JU327752.1 | - | 366 | 85 | ||
| JU470838.1 | - | 515 | 85 | |||
| JV044729.1 | - | 506 | 85 | |||
More Information
Set Membership: |
|

SLC25A10
Genomic Map