Assay Details
| Gene Symbol: FBXL21P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015140960.2 | XP_014996446.1 | 1088 | 115 | ||
| XM_015140961.2 | XP_014996447.1 | 810 | 115 | |||
| XM_015140962.2 | XP_014996448.1 | 2262 | 115 | |||
More Information
Set Membership: |
|

FBXL21P
Genomic Map