Assay Details
| Gene Symbol: POM121C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001110260.4 | XP_001110260.2 | 822 | 108 | ||
| XM_015133306.2 | XP_014988792.2 | 391 | 108 | |||
| XM_028845669.1 | XP_028701502.1 | 822 | 108 | |||
| GenBank mRNA | JU337372.1 | - | 620 | 108 | ||
| JV048853.1 | - | 602 | 108 | |||
| JV609474.1 | - | 689 | 108 | |||
| JV666929.1 | - | 732 | 108 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
This assay is in the 5'UTR region of the XM_015133306.2 transcripts.
This assay is in the 5'UTR region of the XM_015133306.2 transcripts.
Set Membership: |
|

POM121C
Genomic Map