Assay Details
| Gene Symbol: LSM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260573.1 | NP_001247502.1 | 251 | 120 | ||
| XM_028838548.1 | XP_028694381.1 | 215 | 120 | |||
| GenBank mRNA | JU320308.1 | - | 171 | 120 | ||
| JU470631.1 | - | 251 | 120 | |||
| JV577499.1 | - | 56 | 120 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

LSM4
Genomic Map