Assay Details
| Gene Symbol: MSRA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001089547.4 | XP_001089547.4 | 1739 | 111 | ||
| XM_001089777.4 | XP_001089777.3 | 1165 | 111 | |||
| XM_001089893.4 | XP_001089893.3 | 1859 | 111 | |||
| XM_002805259.3 | XP_002805305.3 | 1240 | 111 | |||
| XM_028852339.1 | XP_028708172.1 | 1533 | 111 | |||
| XM_028852340.1 | XP_028708173.1 | 1859 | 111 | |||
| XM_028852341.1 | XP_028708174.1 | 293 | 111 | |||
| XM_028852342.1 | XP_028708175.1 | 1790 | 111 | |||
| XM_028852343.1 | XP_028708176.1 | 1377 | 111 | |||
| XM_028852344.1 | XP_028708177.1 | 1739 | 111 | |||
| XM_028852345.1 | XP_028708178.1 | 1165 | 111 | |||
| XM_028852346.1 | XP_028708179.1 | 1240 | 111 | |||
| XM_028852347.1 | XP_028708180.1 | 1440 | 111 | |||
| XM_028852348.1 | XP_028708181.1 | 1096 | 111 | |||
| XM_028852349.1 | XP_028708182.1 | 1596 | 111 | |||
| XM_028852350.1 | XP_028708183.1 | 1533 | 111 | |||
| XM_028852351.1 | XP_028708184.1 | 430 | 111 | |||
| XM_028852352.1 | XP_028708185.1 | 1440 | 111 | |||
| XM_028852353.1 | XP_028708186.1 | 1096 | 111 | |||
| GenBank mRNA | JU322802.1 | - | 556 | 111 | ||
| JU334597.1 | - | 656 | 111 | |||
| JU470903.1 | - | 488 | 111 | |||
| JV047115.1 | - | 619 | 111 | |||
| JV565127.1 | - | 481 | 111 | |||
More Information
Set Membership: |
|

MSRA
Genomic Map