Assay Details
| Gene Symbol: SYNGR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194830.3 | NP_001181759.1 | 437 | 166 | ||
| XM_001113574.4 | XP_001113574.1 | 614 | 166 | |||
| XM_015124377.2 | XP_014979863.1 | 655 | 166 | |||
| XM_015124378.2 | XP_014979864.1 | 633 | 166 | |||
| XM_028838487.1 | XP_028694320.1 | 382 | 166 | |||
More Information
Set Membership: |
|

SYNGR4
Genomic Map