Assay Details
| Gene Symbol: TIMM13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261039.1 | NP_001247968.1 | 376 | 98 | ||
| GenBank mRNA | JU321556.1 | - | 281 | 98 | ||
| JU473438.1 | - | 315 | 98 | |||
| JV044642.1 | - | 376 | 98 | |||
| JV637998.1 | - | 315 | 98 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TIMM13
Genomic Map