Assay Details
| Gene Symbol: FAM216A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015152779.1 | XP_015008265.1 | 764 | 105 | ||
| GenBank mRNA | JV713220.1 | - | 751 | 105 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM216A| Gene Symbol: FAM216A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015152779.1 | XP_015008265.1 | 764 | 105 | ||
| GenBank mRNA | JV713220.1 | - | 751 | 105 | ||
Set Membership: |
|