Assay Details
Gene Symbol: REM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015149285.2 | XP_015004771.1 | 754 | 66 | ||
XM_015149287.2 | XP_015004773.1 | 562 | 66 | |||
GenBank mRNA | JV044201.1 | - | 708 | 66 | ||
JV649221.1 | - | 409 | 66 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
