Assay Details
| Gene Symbol: IL20RA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001097436.4 | XP_001097436.3 | 660 | 118 | ||
| XM_015137364.2 | XP_014992850.2 | 1134 | 118 | |||
| XM_015137366.2 | XP_014992852.2 | 733 | 118 | |||
| XM_028847153.1 | XP_028702986.1 | 1044 | 118 | |||
| GenBank mRNA | JV717092.1 | - | 669 | 118 | ||
More Information
Set Membership: |
|

IL20RA
Genomic Map