Assay Details
| Gene Symbol: JOSD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266873.1 | NP_001253802.1 | 353 | 74 | ||
| XM_015150318.2 | XP_015005804.1 | 938 | 74 | |||
| XM_015150323.2 | XP_015005809.1 | 422 | 74 | |||
| GenBank mRNA | JU320086.1 | - | 449 | 74 | ||
| JU332160.1 | - | 449 | 74 | |||
| JU332161.1 | - | 449 | 74 | |||
| JU478086.1 | - | 354 | 74 | |||
| JV048862.1 | - | 353 | 74 | |||
More Information
Set Membership: |
|

JOSD1
Genomic Map