Assay Details
| Gene Symbol: ADNP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266818.1 | NP_001253747.1 | 1127 | 90 | ||
| GenBank mRNA | JU333673.1 | - | 410 | 90 | ||
| JU474926.1 | - | 364 | 90 | |||
| JU474927.1 | - | 205 | 90 | |||
| JU474928.1 | - | 410 | 90 | |||
| JV046584.1 | - | 952 | 90 | |||
| JV046585.1 | - | 1127 | 90 | |||
| JV046586.1 | - | 1127 | 90 | |||
More Information
Set Membership: |
|

ADNP
Genomic Map