Assay Details
Gene Symbol: ELP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015118439.2 | XP_014973925.1 | 389 | 67 | ||
XM_028835956.1 | XP_028691789.1 | 389 | 67 | |||
GenBank mRNA | JU483638.1 | - | 118 | 67 | ||
JV611483.1 | - | 325 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
