Assay Details
Gene Symbol: TPGS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260587.2 | NP_001247516.1 | 747 | 79 | ||
XM_015121729.2 | XP_014977215.1 | 577 | 79 | |||
XM_015121730.2 | XP_014977216.1 | 807 | 79 | |||
XM_015121731.2 | XP_014977217.1 | 684 | 79 | |||
XM_015121733.2 | XP_014977219.1 | 937 | 79 | |||
XM_015121734.2 | XP_014977220.1 | 937 | 79 | |||
XM_015121735.2 | XP_014977221.1 | 707 | 79 | |||
GenBank mRNA | JU321611.1 | - | 694 | 79 | ||
JU475069.1 | - | 746 | 79 | |||
JV046690.1 | - | 748 | 79 | |||
JV612369.1 | - | 761 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
