Assay Details
Gene Symbol: MTG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258089.1 | NP_001245018.1 | 847 | 76 | ||
XM_015148638.2 | XP_015004124.2 | 1375 | 76 | |||
XM_015148639.2 | XP_015004125.2 | 1329 | 76 | |||
XM_028827226.1 | XP_028683059.1 | 970 | 76 | |||
GenBank mRNA | JU333931.1 | - | 841 | 76 | ||
JU475115.1 | - | 847 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
