Assay Details
Gene Symbol: MMADHC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265642.1 | NP_001252571.1 | 749 | 118 | ||
XM_015110081.2 | XP_014965567.1 | 925 | 118 | |||
GenBank mRNA | JU320976.1 | - | 796 | 118 | ||
JU475198.1 | - | 793 | 118 | |||
JV613360.1 | - | 791 | 118 | |||
JV667378.1 | - | 749 | 118 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
