Assay Details
Gene Symbol: CHST15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015148395.2 | XP_015003881.1 | 7051 | 109 | ||
XM_015148396.2 | XP_015003882.1 | 1275 | 109 | |||
XM_015148397.2 | XP_015003883.1 | 1115 | 109 | |||
XM_028826975.1 | XP_028682808.1 | 1119 | 109 | |||
XM_028826976.1 | XP_028682809.1 | 1264 | 109 | |||
GenBank mRNA | JU472197.1 | - | 1061 | 109 | ||
JU472198.1 | - | 1061 | 109 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
