Assay Details
Gene Symbol: GMNN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258199.1 | NP_001245128.1 | 397 | 105 | ||
XM_015135643.2 | XP_014991129.2 | 500 | 105 | |||
XM_015135644.2 | XP_014991130.2 | 428 | 105 | |||
XM_015135645.2 | XP_014991131.2 | 477 | 105 | |||
GenBank mRNA | JU323367.1 | - | 289 | 105 | ||
JU473626.1 | - | 397 | 105 | |||
JV043281.1 | - | 296 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() |
