Assay Details
| Gene Symbol: RDH11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257659.1 | NP_001244588.1 | 504 | 78 | ||
| GenBank mRNA | JU334786.1 | - | 504 | 78 | ||
| JU334787.1 | - | 504 | 78 | |||
| JU334788.1 | - | 504 | 78 | |||
| JU334789.1 | - | 504 | 78 | |||
| JU334790.1 | - | 504 | 78 | |||
| JU475824.1 | - | 504 | 78 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RDH11
Genomic Map