Assay Details
| Gene Symbol: MRPL11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261127.1 | NP_001248056.1 | 206 | 78 | ||
| GenBank mRNA | JU335977.1 | - | 206 | 78 | ||
| JU476858.1 | - | 206 | 78 | |||
| JV659137.1 | - | 211 | 78 | |||
More Information
Set Membership: |
|

MRPL11
Genomic Map