Assay Details
| Gene Symbol: GLOD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265952.1 | NP_001252881.1 | 784 | 115 | ||
| XM_028835507.1 | XP_028691340.1 | 715 | 115 | |||
| GenBank mRNA | JU322522.1 | - | 752 | 115 | ||
| JU472652.1 | - | 779 | 115 | |||
| JV044826.1 | - | 784 | 115 | |||
| JV704222.1 | - | 792 | 115 | |||
More Information
Set Membership: |
|

GLOD4
Genomic Map