Assay Details
| Gene Symbol: PCYOX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015112608.2 | XP_014968094.1 | 942 | 148 | ||
| GenBank mRNA | JU334897.1 | - | 898 | 148 | ||
| JU475903.1 | - | 899 | 148 | |||
| JU475904.1 | - | 899 | 148 | |||
| JV047304.1 | - | 916 | 148 | |||
| JV716589.1 | - | 927 | 148 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PCYOX1
Genomic Map